Learn More
Trajan™ Diamond MS CTC PAL Autosampler Syringes
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
Brand: Trajan™ 004770
This item is not returnable.
View return policy
Description
- Lowest syringe carryover
- Reduced non specific sample interaction and increased analyte recovery
- Ultra smooth removable needles with choice of surface treatment.
Specifications
Gas/Liquid chromatography | |
662°F | |
50R-CTC-GT-MS1 | |
50 μL | |
Removable |
Diamond MS Syringe | |
350°C | |
Ea. | |
CTC PAL Autosampler Syringe |