missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Diamond MS CTC PAL Autosampler Syringes

Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.

£108.00

Specifications

For Use With (Application) Gas/Liquid chromatography
Product Type Diamond MS Syringe
Max. Temperature (English) 662°F
Max. Temperature (Metric) 350°C
Quantity Ea.
View More Specs

Products 4
Product Code Brand Volume (Metric) Model Price Quantity & Availability  
Product Code Brand Volume (Metric) Model Price Quantity & Availability  
13157976
View Documents
Trajan™
002770
10 μL 10R-CTC-GT-MS1
£108.00
Each
Please to purchase this item. Need a web account? Register with us today!
13167976
View Documents
Trajan™
003770
25 μL 25R-CTC-GT-MS1
£108.00
Each
Please to purchase this item. Need a web account? Register with us today!
13177976
View Documents
Trajan™
004770
50 μL 50R-CTC-GT-MS1
£108.00
Each
Please to purchase this item. Need a web account? Register with us today!
13187976
View Documents
Trajan™
005770
100 μL 100R-CTC-GT-MS1
£108.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications

Specifications

Gas/Liquid chromatography
662°F
Ea.
Removable
Diamond MS Syringe
350°C
CTC PAL Autosampler Syringe
Videos
SDS
Documents

Documents

Product Certifications