missing translation for 'onlineSavingsMsg'
Learn More

SGE™ Microvolume Manual Syringes

Protection guide and plunger button reinforcement to minimize the risk of bent plungers

Brand:  SGE™ 002135

 View more versions of this product

Product Code. 10696705

  • £270.00 / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Accuracy and reproducibility
  • Borosilicate glass barrel
  • Guided plunger
  • Volume: 10μL
  • Scale length: 54.1mm (2.1 in.)

Specifications

Specifications

Borosilicate Glass
0.63 mm
2 in.
Microvolume Manual Syringe; 10FX-5/0.63C
Cone
23 G
2 in.
50 mm
10 μL
Fixed
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers