Learn More
SGE™ Replacement Needles for Removable Needle Syringes
Replacement needles as supplied with syringe
Brand:  SGE™ 037776
 View more versions of this product
Product Code. 15238593
  • £40.10 / Pack of 2
Explore more special offers
Description
Description
  • For 10μL syringes
  • 50mm (2 in.) exposed length when fitted to syringe
Specifications
Specifications
25 G
0.5 mm
Gas chromatography
1.9 in.
Replacement Needle
50 mm
Removable Needle Syringe (10μL)
0.019 in.
0.5 mm
2 in.
50 mm
2/Pk.
Bevel