Learn More
Cole-Parmer™ SH-200 Series Reciprocating Shakers
Powerful yet quiet, perfect for continuous use applications.
Brand:  Cole-Parmer™ 51901-02
Product Code. 17701288
  • £1730.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Digital speed selection provides consistent mixing
  • Soft-start motor prevents spills and splashes
  • Fully adjustable cradle accommodates most flasks, bottles, and beakers
  • Can be used in incubators system
These linear reciprocating shakers feature digital speed selection from 25 to 250 rpm with a soft start. Four adjustable rubber cushioned horizontal securing bars with quick release handles on the cradle-type platform allow you to fit nearly any size flask, bottle, or beaker. Secure up to twelve 250 mL, nine 500 mL, four 1 L, or two 2 L vessels. Easily set digital timer from 1 to 999 minutes. When timer reaches zero, the shaker automatically stops and sounds an alert. Shaker is capable of continuous use, and the microprocessor maintains speed accuracy over long runs.
Specifications
Specifications
10 kg
230 VAC
50 Hz
45 cm
10.82 in.
0.75 in.
15.35 in.
Large Reciprocating Shaker
1 to 999 rpm
25 to 250 rpm
22 lb.
17.716 in.
33.5 x 33.5 cm (13 x 13 in.)
27.5 cm
20 mm
39 cm
LED
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ SH-200 Series Reciprocating Shakers > Cole-Parmer™ Reciprocating Shakers