Learn More
Memmert™ Shaking Device for Waterbaths With Support Frame
Shaking device with support frame without gable cover & support basket (mandatory)
Brand: Memmert™ WTB24SV1524
This item is not returnable.
View return policy
Description
- Requires gable cover
- Voltage 230 V, 50/60 Hz
- Order the support basket separately
- Includes support frame, shaking frequency 35 - 160 strokes per minute (horizontal back/forth movement)
Specifications
WTB24 | |
Shaking device for waterbaths incl. support frame | |
35-160 rpm |