Learn More
Cole-Parmer™ SI-200 Series Stuart™ Shaking Incubator
Shaker and incubator combination makes this SI-200 Series Stuart™ Shaking Incubator ideal for cell culture procedures.
Brand:  Cole-Parmer™ 79520-18
 View more versions of this product
Product Code. 15847894
  • £7340.00 / Each
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Microprocessor controlled shaking speed ensures accuracy.
  • Forced air circulation ensures uniform temperature inside the chamber.
  • Easily set and view temperature and speed on the digital display.
  • Unique retractable platform offers easy loading and unloading.
  • These shaking incubators provide an orbital shaking motion to ensure even mixing while promoting bacterial growth.
  • All models feature a timer which can be set from 1 second to 9 days.
  • Once the timer has counted down, an alarm will sound and the shaking action will stop while the incubator will continue to run.
  • The retractable platform allows easy access to the samples at the back of the incubator yet securely locks in place during operation.
  • A versatile clamping system holds flasks securely in place (up to 1 L flasks for 51 L models and up to 2 L flasks for 115 L models).
  • Built-in USB connection and dedicated software allow long-term monitoring of the incubator temperature via a PC.
  • Order models 79520-02 or 79520-18 with IQ/OQ (Installation Qualification/Operational Qualification) documentation to help you comply with regulatory agency requirements.
Specifications
Specifications
16 mm
230 V
Large Capacities - 115 L
50 Hz
46.5 x 62.3 x 39.5 cm
39.5 cm
46.5 cm
62.3 cm
Digital
Installation Qualification/Operational Qualification (IQ/OQ)
30 to 300 rpm
60°C (Max.)
140°F max
52.6 x 39 cm
15.6 in.
18.3 in.
24.5 in.
SI600/IQOQ
Cell Culture Applications
1 sec. to 9 days
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ SI-200 Series Stuart™ Shaking Incubator > SI-200 Series Stuart™ Shaking Incubator; Large Capacities - 115 L; Includes: IQ/OQ