Learn More
Epredia™ Shandon™ CytoRich™ Red Collection Fluid
Allow for more accurate evaluation of cytological features with Epredia™ Shandon™ CytoRich™ Red Collection Fluid, which lyses RBCs and solubilizes proteins.
Brand:  Epredia™ B9990800
Code : NEW
Additional Details : CAS Number : 67-63-0 Weight : 4.00000kg Transport : UN number : 1987 Chem class : 3 Pack group : III
 View more versions of this product
Product Code. 15337908
  • £233.00 / 4 L
Estimated Shipment: 29-07-2024
to see stock.
Description
Description
  • Pre-filled containers available
  • Ready-to-use, alcohol-based fluid
  • Compatible with immunohistochemistry
Specifications
Specifications
Collection Fluid
1 gal.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.
  • H371-May cause damage to organs.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : isopropyl alcohol, methyl alcohol, ethylene glycol, formaldehyde, sodium hydroxide, monodoium phosph