Learn More
Epredia™ Shandon™ EA-50 Stain
Clearly differentiate cellular components with pink, green and blue-green hues utilizing Epredia™ Shandon™ EA-50 Stain.
Brand: Epredia™ 6766001
This item is not returnable.
View return policy
Description
- Polychromatic cytoplasmic stain
- Biological stain commission-certified dye
Specifications
1L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Cytological stain EA-50 / EA-65
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Specific target organ toxicity Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H371-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Ethyl alcohol, Methyl alcohol, Isopropylalcohol, Phosphotungstic acid, Eosin-YDye