Learn More
Epredia™ Shandon™ EA-65 Stain
Improve the expression of eosin-stained components with Epredia™ Shandon™ EA-65 Stain, which contains a lower concentration of light green.
Brand:  Epredia™ 6766003
 View more versions of this product
Product Code. 12519996
  • £67.70 / Pack of 2
Explore more special offers
Description
Description
  • Polychromatic cytoplasmic stain
  • Ideal for thicker non-GYN preparations
  • Biological Stain Commission-certified dye
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cellular Components Stain
2 x 1L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Epredia™ Shandon™ EA-65 Stain > 2 x 1L