missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Instant Eosin
Obtain brilliant and consistent staining with Epredia™ Shandon™ Instant Eosin.
Brand: Epredia™ 6765040
135.75 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Stable and precipitate-free; no filtering
- pH-adjusted for maximum stain uptake
- Economical powder stores indefinitely
- Compatible with all alum Hematoxylin stains
Specifications
Alum Hematoxylin Stains | |
Eosin | |
Eosin | |
Liquid |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H302-Harmful if swallowed.
- H412-Harmful to aquatic life with long lasting effects.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction