missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Instant Eosin

Obtain brilliant and consistent staining with Epredia™ Shandon™ Instant Eosin.

£186.00 - £289.00

Specifications

Compatibility Alum Hematoxylin Stains
Product Type Eosin
Solution Type Eosin
Form Liquid
View More Specs

Products 2
Product Code Brand Solution Type Price Quantity & Availability  
Product Code Brand Solution Type Price Quantity & Availability  
12697926
View Documents Promotion Details
Epredia™
6765040
Eosin
£186.00
Pack of 6
Estimated Shipment: 27-12-2024
to see stock.
Add to basket
 
10046318
View Documents Promotion Details
Epredia™
6765540
Eosin
£289.00
Pack of 6
Estimated Shipment: 27-12-2024
to see stock.
Add to basket
 
Description

Description

  • Stable and precipitate-free; no filtering
  • pH-adjusted for maximum stain uptake
  • Economical powder stores indefinitely
  • Compatible with all alum Hematoxylin stains
Specifications

Specifications

Alum Hematoxylin Stains
Eosin
Eosin
Liquid
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Product Certifications