missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Straight Metzenbaum Scissors, Straight, standard, 5.5 in. (14.0cm)
Epredia™ Shandon Straight Metzenbaum Scissors are available in Premium and Standard grades.
£77.62
Product Code | Brand | Grade | Length (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Grade | Length (English) | Price | Quantity & Availability | ||||
15624454
|
Epredia™
79201 |
Premium | 5.5 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Consistent Quality- Properly hardened, tempered and passivated
- Electronically polished and cleaned for a high-quality finish
- Patterns, dimensions and finish held to exacting standards
- Guaranteed to be free from defects in material and workmanship
Specifications
Metzenbaum Scissors | |
Straight |