Learn More
Epredia™ Cassette 1 Slotted Tissue Cassettes
Hold tissue specimens during processing and embedding with Epredia™ Cassette 1 slotted tissue cassettes.
£100.47
Specifications
Pore Style | Slotted Pore |
---|---|
Writing Face Angle | 30° |
Product Type | Tissue Cassette |
Product Code | Brand | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | |||||
15327260
|
Epredia™
1000961 |
Blue | N/A | ||||||
Description
- Acetal polymer construction
- 30° writing face
- Small slot-style pores
- Supplied in box of 500 cassettes with attached breakaway lids (not available in Tube Packs)
Specifications
Slotted Pore | |
30° | |
Tissue Cassette |