missing translation for 'onlineSavingsMsg'
Learn More

Sheep anti-Rabbit IgG (H+L) Secondary Antibody, Biotin, Novus Biologicals™

Sheep Polyclonal Antibody

Marca:  Bio-Techne NB120-6794

190.40 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.


Código de producto. 18169679

  • £238.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

IgG (H+L)
Polyclonal
Western Blot 1:1000 - 1:10000, ELISA 1:300000
Sheep
Multi-step
RUO
Secondary
Rabbit
Purified
Western Blot, ELISA
Biotin
IGHG
Rabbit IgG whole molecule
1 mg
Adaptive Immunity, Immunology
Rabbit IgG Heavy and Light
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

One moment while we fetch your results.
Certificados
Ofertas especiales

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sheep anti-Rabbit IgG (H+L) Secondary Antibody, Biotin, Novus Biologicals™ > 1mg; Biotin

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only