Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Sulfide
For gas detection
Brand:  Dräger™ 8101991
 View more versions of this product
Product Code. 10010022
  • £134.00 / Pack of 10
Explore more special offers
Description
Description
Note: Extension of measuring range listed is possible for some detector tubes. Call Fisher Customer Service for more information.
Specifications
Specifications
Hydrogen Sulfide 0.2/b
0.2 to 6ppm
10
59°F to 104°F
20mg/L
1
55 sec.
15° to 40°C