Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Sulfide
For gas detection
Brand:  Dräger™ 8101961
Code : 50
Additional Details : Weight : 0.08460kg
 View more versions of this product
Product Code. 10473172
  • £184.00 / Pack of 10
Estimated Shipment: 13-09-2024
to see stock.
Description
Description
Note: Extension of measuring range listed is possible for some detector tubes. Call Fisher Customer Service for more information.
Specifications
Specifications
Hydrogen Sulfide 2/b
2 to 60ppm
10
32°F to 104°F
20mg/L
1
30 sec.
0°C to 40°C
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • drager tubes hydrogen sulphide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.