Learn More
Bohlender™ SICCO™ Glove Box Duo 2 with Transfer Chamber
Glove Box Duo 2 with Transfer Chamber with panels made of acrylic glass and base panel made of polyethylene.
Brand:  Bohlender™ V1710-08
Product Code. 17167674
  • £12720.00 / Each
Explore more special offers
Description
Description
  • Aluminum frame with panels made of acrylic glass, base panel made of polyethylene
  • Front and back side with one door each, two pairs of opposite glove ports each with one pair of natural rubber gloves size 9
  • Two cable lead-ins (left bottom side)
  • Transfer chamber (right) with two doors, external door with latch to prevent accidental opening
  • Tray extends into the workspace with opening of the internal door for easy removal of inserted products and is only operable from one workstation side (front right-hand)
  • Work space and transfer chamber include two hose connectors for gas-filling each and appropriate tubing (2 x 5 m tubing for gas-filling made of PVC and 2 x 5 m aeration tubing made of PE)
  • Robust and light-weight aluminum construction
  • Superb all-round visibility
  • Non-slip rubber feet
  • Cable lead-ins for electric devices
  • Large front opening
  • Connectors for gas-filling and aeration including appropriate tubing
  • Transfer chamber for inserting further products
Specifications
Specifications
Acrylic Glass
33 in.
845 mm
26 in.
665 mm
66 lb.
47 in.
1200 mm
35 in.
890 mm
28 in.
710 mm
475 L
30 kg
33 in.
850 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Bohlender™ SICCO™ Glove Box Duo 2 with Transfer Chamber > Glove Box Duo 2 with Transfer Chamber; Material: Acrylic Glass; Depth (Metric) Exterior: 890 mm; Height (Metric) Exterior: 710 mm; Width (Metric) Exterior: 1200 mm; Weight (Metric): 30 kg