Learn More
SymLine™ SignalBox
This built-in signal box monitors up to five filling levels at once, making it ideal for integration into lab furniture
Brand: SymLine™ 106548
469.47 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
- Clever way to change a disposal container without interrupting the inflow.
- LEDs and acoustic buzzer warn you in due time before the filling level of your collection containers reaches a critical height.
- When container is changed, the alert can be muted via the new touchpad.
- Includes touchless alarm-resetting fields for up to five containers, up to five switch inputs and up to five switch outputs for peripheral units, and power supply cable with open end and clamp.
Specifications
2-channel | |
Safety Waste Cap and Electronic Level Controllers | |
36 x 139 x 74 mm |
Liquid Spill Alarm | |
SignalBox2, Power Supply Cable with Open End and Clamp | |
EU-style |