Learn More
SymLine™ SignalBox
This built-in signal box monitors up to five filling levels at once, making it ideal for integration into lab furniture
£355.00 - £674.00
Specifications
Type | Liquid Spill Alarm |
---|---|
For Use With (Equipment) | Safety Waste Cap and Electronic Level Controllers |
Product Code | Brand | Design | Plug Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Design | Plug Type | Price | Quantity & Availability | ||||||
16583532
|
SymLine™
106548 |
2-channel | EU-style |
£643.00
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||||
17307419
|
SymLine™
106582 |
2-channel | UK-style | N/A | N/A | N/A | |||||
17317419
|
SymLine™
106583 |
2-channel | US-style | N/A | N/A | N/A | |||||
18686468
|
SymLine™
106735 |
1-channel | EU, UK, US |
£674.00
1 sheet |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
18666498
|
SymLine™
106741 |
5-channel | EU, UK, US |
£355.00
1 sheet |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||||
Description
- Clever way to change a disposal container without interrupting the inflow.
- LEDs and acoustic buzzer warn you in due time before the filling level of your collection containers reaches a critical height.
- When container is changed, the alert can be muted via the new touchpad.
- Includes touchless alarm-resetting fields for up to five containers, up to five switch inputs and up to five switch outputs for peripheral units, and power supply cable with open end and clamp.
Specifications
Liquid Spill Alarm | |
Safety Waste Cap and Electronic Level Controllers |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
SymLine™ SignalBox