Learn More
BRAND™ SILBERBRAND Automatic Burette with Lateral Safety Stopcock
SILBERBRAND Automatic Burette includes a lateral safety stopcock and black Schellbach stripe.
Brand: BRAND™ 21911
188.95 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Calibrated to deliver (TD, Ex)
- With needle-valve stopcock with PTFE spindle
- Schellbach stripe
- Automatic zeroing
Specifications
10 mL | |
2000 mL bottle | |
12 months |
0.02 mL | |
Lateral safety stopcock |