missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ SilberBrand™ Boro 3.3 Compact-Burette Tube

Economy class B burette with removable PTFE stopcock and PP tip

Brand:  BRAND™ 10014

 View more versions of this product

Product Code. 17480124

  • £85.40 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Error limits according to class B, DIN EN ISO 385
  • Calibrated to deliver (TD, Ex)
  • TRUSTED_SUSTAINABILITY
    Specifications

    Specifications

    Error Limit: 0.10 mL
    Borosilicate Glass
    2
    50 mL
    Class B
    50 mL
    0.1 mL
    Removable PTFE Stopcock and PP Tip
    Compact Buret
    DIN EN ISO 385
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.