Learn More
Brand™ Silberbrand™ Eterna™ AR-GLAS™ Class B Measuring Pipets
Manufactured from AR-GLAS™ soda lime glass, this Class B Measuring Pipet is ideal in most research, scientific, and other laboratory environments.
Brand: Brand™ 27078
43.71 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured from AR-GLAS™ Soda lime glass
- Type 3, with zero value at the top
- Amber stain marking
- Class B
- Compliant with DIN EN ISO 835 standards
- Calibrated to deliver (TD, Ex)
- Includes enlarged suction tube for cotton plugs
Specifications
Yes | |
±0.15 mL | |
20 mL |
0.1 mL | |
Green | |
360 mm |