Learn More
Product Code | Brand | Volume (Metric) | Graduations | Tolerance | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Graduations | Tolerance | Color | Price | Quantity & Availability | |||||
11778158
|
Brand™
27072 |
2 mL | 0.01 mL | ±0.015 mL | White |
£67.70
Pack of 12 |
In Stock
Log in to see stock. |
|||||
12055302
|
Brand™
27069 |
5 mL | 0.01 mL | ±0.008 mL | 2 x Green | N/A | ||||||
11853970
|
Brand™
27073 |
2 mL | 0.02 mL | ±0.015 mL | Black |
£56.95
Pack of 12 |
In Stock
Log in to see stock. |
|||||
12065302
|
Brand™
27070 |
1 mL | 0.01 mL | ±0.010 mL | Yellow |
£54.55
Pack of 12 |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
12075302
|
Brand™
27071 |
1 mL | 0.1 mL | ±0.010 mL | Red | N/A | ||||||
11873970
|
Brand™
27076 |
5 mL | 0.1 mL | ±0.05 mL | Blue |
£56.35
Pack of 12 |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
11863970
|
Brand™
27075 |
5 mL | 0.05 mL | ±0.05 mL | Red |
£61.80
Pack of 12 |
In Stock
Log in to see stock. |
|||||
11883970
|
Brand™
27078 |
20 mL | 0.1 mL | ±0.15 mL | Green |
£54.10
Pack of 6 |
In Stock
Log in to see stock. |
|||||
11893970
|
Brand™
27079 |
25 mL | 0.1 mL | ±0.15 mL | White |
£62.75
Pack of 6 |
In Stock
Log in to see stock. |
|||||
11788158
|
Brand™
27077 |
10 mL | 0.1 mL | ±0.08 mL | Orange |
£101.00
Pack of 12 |
Estimated Shipment: 29-04-2025
Log in to see stock. |
|||||
Description
- Manufactured from AR-GLAS™ Soda lime glass.
- Type 3, with zero value at the top.
- Amber stain marking.
- Class B.
- Compliant with DIN EN ISO 835 standards.
- Calibrated to deliver (TD, Ex).
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Brand™ Silberbrand™ Eterna™ AR-GLAS™ Class B Measuring Pipets