Learn More
Ambion™ Silencer™ Custom Designed siRNA
Silencer™ custom siRNAs (unmodified 21-bp duplexes) are cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types.
Brand:  Ambion™ 4399670
 View more versions of this product
Product Code. 15511471
Description
Description
  • Cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types
  • Use the GeneAssist™ Custom siRNA Builder to design and order a custom siRNA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at or below -20°C. Do not store in a frost-free freezer. (Dried oligonucleotides are shipped at ambient temperature.) Additional Materials Included: 1.75 mL Nuclease-free Water.
Knocking down gene expression in order to study protein function in a wide range of cell types
Custom Designed siRNA
5 nmol
Upon receipt, store in a non-frost-free freezer at or below -20°C
GeneAssist - Custom Designed Silencer™ siRNA
Silencer™
Standard
Ambient
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ambion™ Silencer™ Custom Designed siRNA > GeneAssist - Custom Designed Silencer™ siRNA, Standard, 5 nmol