missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambion™ Silencer™ Custom Designed siRNA
Silencer™ custom siRNAs (unmodified 21-bp duplexes) are cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types.
£143.00 - £466.00
Specifications
Content And Storage | Store at or below -20°C. Do not store in a frost-free freezer. (Dried oligonucleotides are shipped at ambient temperature.) Additional Materials Included: 1.75 mL Nuclease-free Water. |
---|---|
Description | GeneAssist - Custom Designed Silencer™ siRNA |
Product Line | Silencer™ |
Product Type | Custom Designed siRNA |
Shipping Condition | Ambient |
Product Code | Brand | Purity | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Purity | Quantity | Price | Quantity & Availability | ||||
15551054
|
Ambion™
4399670 |
Standard | 5 nmol |
£188.00
5nmol |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15561054
|
Ambion™
4399671 |
Standard | 20 nmol |
£258.00
20nmol |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15571054
|
Ambion™
4399672 |
Standard | 40 nmol |
£328.00
40nmol |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15581054
|
Ambion™
4399673 |
HPLC | 20 nmol |
£394.00
20nmol |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15591054
|
Ambion™
4399674 |
HPLC | 40 nmol |
£466.00
40nmol |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15511471
|
Ambion™
4399670 |
Standard | 5 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15521471
|
Ambion™
4399671 |
Standard | 20 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15541471
|
Ambion™
4399673 |
HPLC | 20 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15531471
|
Ambion™
4399672 |
Standard | 40 nmol |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types
- Use the GeneAssist™ Custom siRNA Builder to design and order a custom siRNA
Specifications
Store at or below -20°C. Do not store in a frost-free freezer. (Dried oligonucleotides are shipped at ambient temperature.) Additional Materials Included: 1.75 mL Nuclease-free Water. | |
Silencer™ | |
Ambient |
GeneAssist - Custom Designed Silencer™ siRNA | |
Custom Designed siRNA | |
Upon receipt, store in a non-frost-free freezer at or below -20°C |
For Research Use Only. Not for use in diagnostic procedures.