Learn More
Ambion™ Silencer™ Custom Designed siRNA
Silencer™ custom siRNAs (unmodified 21-bp duplexes) are cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types.
Brand: Ambion™ 4399671
Description
- Cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types
- Use the GeneAssist™ Custom siRNA Builder to design and order a custom siRNA

Specifications
Store at or below -20°C. Do not store in a frost-free freezer. (Dried oligonucleotides are shipped at ambient temperature.) Additional Materials Included: 1.75 mL Nuclease-free Water. | |
Knocking down gene expression in order to study protein function in a wide range of cell types | |
Custom Designed siRNA | |
20 nmol | |
Upon receipt, store in a non-frost-free freezer at or below -20°C |
GeneAssist - Custom Designed Silencer™ siRNA | |
Silencer™ | |
Standard | |
Ambient |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.