Learn More
Thermo Scientific™ Cyto-Plex™ Carboxylated Particles
Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.
Brand: Thermo Scientific™ FM5CR08
Alert:
Contains sodium azide.
Description
Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.
Suspension array analysis is a powerful technique enabling the simultaneous detection and quantitation of multiple analytes with a single sample. Using color coded microspheres with the appropriate dyes, these tests can now be carried out using commonly available flow cytometers.- Cyto-Plex™ microspheres consist of a highly uniform polystyrene particle with fluorescent intensities which are completely separated from each other
- Carboxylated particles provide different sizes and levels of fluorescent intensities for analysis of multiple analytes
- Use of a single diameter particle for all dye levels saves time by only requiring the development and optimization of one bead chemistry; multiple diameters can be combined to increase the number of analytes measured in one test
- High-density binding sites and low non-specific binding enable coupling with a wide variety of antibodies, nucleic acids and other biomolecules
- Microspheres have a maximum emission at 700 nm and can be excited with either 488 nm or 633 nm lasers
- Emission can be collected in either the PE-Cy5 or APC channels
- Since there is little or no emission in the FITC and PE channels, probes utilizing either of these dyes can be effectively used as reporters
Recommended for:
- Open platform microspheres for suspension array analysis in flow cytometry

Specifications
Sodium Azide Preservative, 0.05% Tween-20 | |
Polystyrene microspheres in water | |
Red | |
Carboxylated Particle | |
Material Safety Data Sheet (MSDS) and Package Insert Sheet | |
1.06g/cm³ | |
Product stable for a minimum of 12 months | |
≤2% CV | |
5 μm |
Refrigerate when not in use; do not freeze. Store upright and keep bottle tightly sealed. Mix product by gentle inversion by hand or vortex mixer. | |
Cyto-Plex Carboxylated Particles, 5µm | |
Polystyrene with Copolymer Grafted Surface | |
Firefli™ Fluorescent Red (635/700nm) | |
Level 8 | |
1 mL | |
Carboxylate-Modified | |
1 or 5 mL |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
273728_1125 | Certificate of Analysis | 28/01/2025 | FM5CR08 |
286283_1226 | Certificate of Analysis | 25/01/2025 | FM5CR08 |
222543 | Certificate of Analysis | 28/01/2022 | FM5CR08 |
224939 | Certificate of Analysis | 28/01/2022 | FM5CR08 |
242173 | Certificate of Analysis | 28/01/2022 | FM5CR08 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.