Learn More
Ambion™ Silencer™ Custom Designed siRNA
Silencer™ custom siRNAs (unmodified 21-bp duplexes) are cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types.
Brand: Ambion™ 4399674
326.29 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
- Cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types
- Use the GeneAssist™ Custom siRNA Builder to design and order a custom siRNA

Specifications
Store at or below -20°C. Do not store in a frost-free freezer. (Dried oligonucleotides are shipped at ambient temperature.) Additional Materials Included: 1.75 mL Nuclease-free Water. | |
siRNA | |
Custom Designed siRNA | |
40 nmol | |
Upon receipt, store in a non-frost-free freezer at or below -20°C |
GeneAssist - Custom Designed Silencer™ siRNA | |
Silencer™ | |
HPLC | |
Ambient |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.