Learn More
Ambion™ Silencer™ Select Custom Designed siRNA
Silencer™ custom siRNAs (unmodified 21-bp duplexes) are cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types.
Brand:  Ambion™ 4399667
256.61 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
 View more versions of this product
Product Code. 15521054
  • £383.00 / 40nmol
Explore more special offers
Description
Description
  • Cost-effective RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types
  • Use the GeneAssist™ Custom siRNA Builder to design and order a custom siRNA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Contains dried siRNA and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below -20°C.
Knocking down gene expression in order to study protein function in a wide range of cell types
Custom Designed siRNA
40 nmol
Upon receipt, store in a non-frost-free freezer at or below -20°C
GeneAssist - Custom Designed Silencer™ Select siRNA
Silencer™ Select
Standard
Ambient