missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Silencer™ Select Pre-Designed siRNA

Silencer™ Select Pre-Designed siRNA

Brand:  Ambion™ 4390822

423.28 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.


 View more versions of this product

Product Code. 10299774

  • £572.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

The Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry. A choice of Silencer™ Select siRNAs are available—Pre-Designed or Validated. Silencer™ Select Pre-Designed siRNAs are guaranteed-to-silence based on their proven design while the Silencer™ Select Validated siRNAs are functionally tested to reduce target gene expression. Sets of Silencer™ Select siRNA Libraries are also available in tubes or plate for high throughput screening. The Silencer™ Select Positive and Negative Control RNAs should be included in every siRNA experiment.

The benefits of using Silencer™ Select siRNAs include:
  • Potent—up to 100-fold more potent than currently available siRNAs and fewer off-target effects
  • Most Specific—LNA™ chemical modifications reduce off-target effects by up to 90%
  • Reliable—demonstrated improvements in consistency and reliability of phenotypic results
  • Guaranteed—100% guaranteed to silence, the best guarantee in the industry

  • Note: Silencer™ Pre-Designed & Validated siRNAs are designed using our first-generation algorithm and DO NOT include any chemical modifications. They are recommended for completing in vitro RNAi studies that were started using Silencer™ siRNAs. For new studies we recommend Silencer Select siRNAs.

    Approximate Turnaround Times*
  • Standard purification: 4 business days
  • HPLC purification: 6 business days (20 and 40 nmol); 15 business days (250 nmol)
  • In Vivo Ready: 15 business days (250 nmol)
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Contains dried siRNA at (40 nmol) and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below –20 °.
    siRNA
    Silencer™ Select validated siRNA
    40 nmol
    Silencer™ Select Validated siRNA, 40nmol, HPLC Purity, –20°C Storage, Potent, Most Specific, Reliable, Guaranteed, Incorporate the Latest Improvements in siRNA design, Off-Target Effect Prediction Algorithms and Chemistry
    Silencer™ Select, Ambion™
    HPLC
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    One moment while we fetch your results.
    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Ambion™ Silencer™ Select Pre-Designed siRNA > 40 nmol

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.