missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Silica gel, for column chromatography, 0.035-0.070mm, 60Å
For column chromatography.
£91.35 - £1366.80
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10273542
|
Thermo Scientific Chemicals
240360050 |
5 kg |
£381.90
5kg |
In Stock
Log in to see stock. |
|||||
10318090
|
Thermo Scientific Chemicals
240360010 |
1 kg |
£91.35
1kg |
In Stock
Log in to see stock. |
|||||
10446042
|
Thermo Scientific Chemicals
240360300 |
30 kg |
£1366.80
30kg |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For column chromatographyChemical Identifiers
7631-86-9 |
Specifications
60 Å | |
Silica Gel | |
Column Chromatography |
Plastic Drum | |
Silica Gel | |
7631-86-9 |
Safety and Handling
Product Identifier
- Silica gel
RUO – Research Use Only