Learn More
Macherey-Nagel™ Silica Glass Plates
Brand:  Macherey-Nagel™ 811051
Code : 50
Additional Details : Weight : 0.01000kg
Product Code. 10002831
  • £452.00 / Pack of 50
Estimated Shipment: 18-09-2024
to see stock.
Add to basket
Specifications
Silica 60
60 Å
Glass Plate
∼500 m2/g
10 x 20 cm
2 to 10 μm
50 Pk.
0.75 mL/g
0.20 mm