missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Silica Glass Plates

HPTLC glass plates in different plate sizes for many different applications. Silica gel with octadecyl (C18, ODS, RP18) modification.

Brand:  Macherey-Nagel™ 811052

Code : 16

Additional Details : Weight : 0.01000kg

Product Code. 10471662

  • £332.30 / Pack of 25
Estimated Shipment: 11-09-2024
to see stock.
Add to basket

Description

Description

HPTLC Glass plates, modified silica gel layer, Nano-SIL C18-100

  • Base material: Glass plates coated in silica gel
  • Mode: Reverse Phase
  • Layer Hardness: Standard, Soft

Adsorbent:

  • Silica gel with octadecyl (C18, ODS, RP18) modification
  • Phase Nano-SIL C18-100
  • UV Indicator: No
  • Particle Type: Fully porous particles (FPP)
  • Particle Shape: Irregular
  • Particle Size: 2-10 μm
  • Pore Size: 60 Å
  • Pore Volume: 0.75 mL/g
  • Binder: Yes

Indicators:

This plate can be used different UV indicators - manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm); special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm).

Binder:

Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents

Recommended Applications:

Alkaloids, Amino acids, Barbiturates, Drugs, Flavonoids, Indole derivatives, Optical brighteners, Peptides, Phenols, Polycyclic aromatic hydrocarbons (PAH), Preservatives, Reversed phase (RP), Steroids, Vegetable oils (identification of fatty oils acc. to EP)

 

General TLC Plate Features from Macherey-Nagel:

  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
  • The plate can be easily labelled for clear identification
  • Glass plates are robust, heat proof and chemically resistant to all common mobile phases and visualization reagents.
  • Increased separation efficiency due to an optimized particle size distribution
  • A dust free plate allows convenient handling and avoids silica contamination of the TLC chamber
Specifications

Specifications

Silica 60
60 Å
Glass Plate
∼500 m2/g
10 x 10 cm
2 to 10 μm
25 Pk.
0.75 mL/g
0.20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers