missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Standard SIL G Silica Layers on Glass Plates

Product Code. 10609011 Shop All Macherey-Nagel Products
Click to view available options
Dimensions (L x W):
50 x 100 mm
50 x 200 mm
50x200mm
100 x 100 mm
100 x 200 mm
200 x 200 mm
200 x 200 cm
Quantity:
100 Pk.
12 Pk.
15 Pk.
20 Pk.
25 Pk.
25/Pk.
50 Pk.
Thickness (Metric):
0.25 mm
0.5 mm
1 mm
2 mm
Unit Size:
Pack of 100
Pack of 12
Pack of 15
Pack of 20
Pack of 25
Pack of 50
Fluorescent Indicator:
UV254
UV254 + UV366
This item is not returnable. View return policy

Product Code. 10609011

Brand: Macherey-Nagel™ 809061

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G silica layers on glass plates come in a variety of plate sizes and layer thicknesses. Select a fluorescent indicator and choose from a variety of pack sizes.

Glass TLC plates, SIL G/UV 254.
  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
Absorbent
  • Silica 60
  • specific surface (BET) ∼ 500 m2/g
  • Mean pore size 60 Å
  • Specific pore volume 0.75 mL/g,
  • Particle size 5–17 μm
  • Standard grade
Thickness
  • For analytical plates 0.25 mm
  • For preparative plates 0.5 and 1 mm
  • For 2 mm preparative layers a slightly coarser material is used.
Inicators
  • Manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
  • Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm)
Binders
  • Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents

Specifications

Material Silica 60
Particle Size 5 to 17 μm
Pore Size 60 Å
Quantity 15 Pk.
Type Glass Plate
Pore Volume 0.75 mL/g
Specific Surface ∼500 m2/g
Thickness (Metric) 1 mm
Dimensions (L x W) 200 x 200 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.