missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon, 99+%, powder, -325 mesh, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 2355.0°C |
Melting Point | 1410.0°C |
Color | Gray |
Physical Form | Fine Powder |
Quantity | 500 g |
Assay Percent Range | 99+% |
Merck Index | 15, 8630 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in molten alklai oxides,soluble in a mixture of nitric acid and hydrofluor,alkalis,insoluble in nitric acid and hydrochloric acid |
Formula Weight | 28.09 |
Percent Purity | ≥99% |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0465280 | Certificate of Analysis | 12/08/2024 | 201565000 |
A0459234 | Certificate of Analysis | 10/01/2024 | 201560100,201565000,201561000 |
A0374282 | Certificate of Analysis | 14/12/2022 | 201560100,201565000,201561000 |
A0384795 | Certificate of Analysis | 13/12/2022 | 201560100,201565000,201561000 |
A0440843 | Certificate of Analysis | 06/04/2022 | 201560100,201565000,201561000 |
Safety and Handling

Product Identifier
- Silicon powder
- Warning
- Flammable solid Category 2
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Silicon, 99+%, powder, -325 mesh, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction