Learn More
Silicon, 99+%, powder, -325 mesh, Thermo Scientific Chemicals
£13.37 - £120.60
Chemical Identifiers
CAS | 7440-21-3 |
---|---|
Molecular Formula | Si |
Molecular Weight (g/mol) | 28.09 |
MDL Number | MFCD00085311 |
InChI Key | XUIMIQQOPSSXEZ-UHFFFAOYSA-N |
Synonym | metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 |
PubChem CID | 5461123 |
ChEBI | CHEBI:27573 |
IUPAC Name | silicon |
SMILES | [Si] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10092760
|
Thermo Scientific Acros
201560100 |
10 g | Glass Bottle |
£13.37
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10626412
|
Thermo Scientific Acros
201561000 |
100 g | Plastic bottle |
£37.63
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10732125
|
Thermo Scientific Acros
201565000 |
500 g | Plastic bottle |
£120.60
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-21-3 | |
28.09 | |
XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
5461123 | |
silicon |
Si | |
MFCD00085311 | |
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
CHEBI:27573 | |
[Si] |
Specifications
2355.0°C | |
7440-21-3 | |
Fine Powder | |
Si | |
15, 8630 | |
Solubility in water: insoluble. Other solubilities: soluble in molten alklai oxides, soluble in a mixture of nitric acid and hydrofluor, alkalis, insoluble in nitric acid and hydrochloric acid | |
[Si] | |
28.09 | |
CHEBI:27573 | |
≥99% |
1410.0°C | |
Gray | |
99+% | |
MFCD00085311 | |
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
silicon | |
5461123 | |
28.09 | |
Silicon, -325 mesh |
Safety and Handling
GHS H Statement:
Flammable solid.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- Silicon powder
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.