missing translation for 'onlineSavingsMsg'
Learn More
Silicon carbide, beta-phase, 99% (metals basis), Thermo Scientific Chemicals Product Code.: 11387757

Silicon carbide, beta-phase, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11387757
2 kg
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11387757

Brand: Thermo Scientific Alfa Aesar 014165.A3

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

S.A. 11.5 m2/g

Used in abrasives, in polishing and grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are as light emitting diodes and sensors. It is utilized for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic. It compositely plays an important role in the crack-healing behavior.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicideShow MoreShow Less
PubChem CID 9863
ChEBI CHEBI:29390
IUPAC Name methanidylidynesilylium
SMILES [C-]#[Si+]

Specifications

Melting Point 2700°C
Color Black to Gray
Physical Form Powder
Quantity 2 kg
Assay Percent Range 99.8% (metals basis)
Solubility Information Soluble in molten sodium hydroxide,potassium hydroxide and in molten iron.
Formula Weight 40.1
Odor Odorless
Chemical Name or Material Silicon carbide, beta-phase

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN10B008 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014165.22,014165.A3,014165.36
Lot NumberR08F018 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014165.22,014165.A3,014165.36
Lot NumberW10A008 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014165.22,014165.A3,014165.36
Lot NumberT29A010 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014165.22,014165.A3,014165.36
Lot NumberK29L38 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number014165.22,014165.A3,014165.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicon carbide, beta-phase, 99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.