missing translation for 'onlineSavingsMsg'
Learn More
Silicon carbide powder, superfine, 600 grit, Thermo Scientific Chemicals Product Code.: 11490054

Silicon carbide powder, superfine, 600 grit, Thermo Scientific Chemicals

Product Code. 11490054
2500 g
Click to view available options
Quantity:
10,000 g
2500 g
500 g
This item is not returnable. View return policy

Product Code. 11490054

Brand: Thermo Scientific Alfa Aesar A13561.0E

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Silicon carbide powder is used for its hardness in abrasive machining processes such as grinding, honing, water-jet cutting and sandblasting. And is also used as a support and shelving material in high temperature kilns such as for firing ceramics, glass fusing, or glass casting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon carbide powder is used for its hardness in abrasive machining processes such as grinding, honing, water-jet cutting and sandblasting. And is also used as a support and shelving material in high temperature kilns such as for firing ceramics, glass fusing, or glass casting.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicideShow MoreShow Less
PubChem CID 9863
ChEBI CHEBI:29390
SMILES [C-]#[Si+]

Specifications

Quantity 2500 g
Solubility Information Insoluble in water.
IUPAC Name methanidylidynesilanylium
Formula Weight 40.1
Odor Odorless
Density 3.16
Chemical Name or Material Silicon carbide powder, superfine, 600 grit

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10245660 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA13561.0C,A13561.36,A13561.0E
Lot Number10187469 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog NumberA13561.0C,A13561.36,A13561.0E
Lot Number10240101 Certificate TypeCertificate of Analysis Date13/04/2022 Catalog NumberA13561.0C,A13561.36,A13561.0E
Lot Number10235933 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13561.0C,A13561.36,A13561.0E
Lot Number10189247 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13561.0C,A13561.36,A13561.0E

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicon carbide powder, superfine, 600 grit, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.