missing translation for 'onlineSavingsMsg'
Learn More

Silicon carbide powder, superfine, 600 grit, Thermo Scientific Chemicals

£58.89 - £475.37

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
PubChem CID 9863
ChEBI CHEBI:29390
SMILES [C-]#[Si+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11480054
View Documents
Thermo Scientific Alfa Aesar
A13561.36
500 g
£58.89
500g
Please to purchase this item. Need a web account? Register with us today!
11490054
View Documents
Thermo Scientific Alfa Aesar
A13561.0E
2500 g
£157.79
2500g
Please to purchase this item. Need a web account? Register with us today!
11400064
View Documents
Thermo Scientific Alfa Aesar
A13561.0C
10,000 g
£475.37
10000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Silicon carbide powder is used for its hardness in abrasive machining processes such as grinding, honing, water-jet cutting and sandblasting. And is also used as a support and shelving material in high temperature kilns such as for firing ceramics, glass fusing, or glass casting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Silicon carbide powder is used for its hardness in abrasive machining processes such as grinding, honing, water-jet cutting and sandblasting. And is also used as a support and shelving material in high temperature kilns such as for firing ceramics, glass fusing, or glass casting.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

409-21-2
40.10
HBMJWWWQQXIZIP-UHFFFAOYSA-N
9863
[C-]#[Si+]
CSi
MFCD00049531
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
CHEBI:29390

Specifications

409-21-2
MFCD00049531
Insoluble in water.
[C-]#[Si+]
40.10
CHEBI:29390
Odorless
Silicon carbide powder, superfine, 600 grit
CSi
silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
HBMJWWWQQXIZIP-UHFFFAOYSA-N
methanidylidynesilanylium
9863
40.1
3.16
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 206-991-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only