Learn More
Silicon carbide whiskers, 99% (metals basis), Thermo Scientific Chemicals
Description
Used in abrasives, polishing, grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are the light-emitting diodes and sensors. It is utilitarian for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic, compositely play an important role in crack-healing behavior.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 2700°C |
Color | Black to Gray |
Quantity | 50 g |
Assay Percent Range | 99% (metals basis) |
Solubility Information | Soluble in molten alkalis (NaOH,KOH) and molten iron. Insoluble in water. |
Formula Weight | 40.1 |
Odor | Odorless |
Density | 3.2 g/cm3 at 20°C |
Chemical Name or Material | Silicon carbide whiskers |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T30A017 | Certificate of Analysis | 04/01/2024 | 038787.09,038787.18,038787.30 |
K08M09 | Certificate of Analysis | 08/12/2021 | 038787.09,038787.18,038787.30 |
S13E049 | Certificate of Analysis | 08/12/2021 | 038787.09,038787.18,038787.30 |
S17C012 | Certificate of Analysis | 08/12/2021 | 038787.09,038787.18,038787.30 |
C28X048 | Certificate of Analysis | 08/12/2021 | 038787.09,038787.18,038787.30 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.