missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silicon diOxide, purum p.a., Acid purified, Honeywell Fluka™
Purum p.a., acid purified
£49.35 - £388.65
Chemical Identifiers
CAS | 60676-86-0 |
---|---|
Molecular Formula | O2Si |
Molecular Weight (g/mol) | 60.083 |
MDL Number | MFCD00011232 |
InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
Synonym | silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous |
PubChem CID | 24261 |
ChEBI | CHEBI:30563 |
IUPAC Name | dioxosilane |
SMILES | O=[Si]=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15617340
|
Honeywell-Fluka
84880-500G |
500 g | N/A | ||||||
15607340
|
Honeywell-Fluka
84880-1KG |
1 kg |
£102.39
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15627340
|
Honeywell-Fluka
84880-5KG |
5 kg |
£388.65
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Chemical Identifiers
60676-86-0 | |
60.083 | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
24261 | |
dioxosilane |
O2Si | |
MFCD00011232 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
CHEBI:30563 | |
O=[Si]=O |
Specifications
60676-86-0 | |
SiO2 | |
NONH for all modes of transport | |
VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
dioxosilane | |
24261 | |
60.08g/mol | |
Silicon dioxide |
O2Si | |
MFCD00011232 | |
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
O=[Si]=O | |
60.083 | |
CHEBI:30563 | |
2.6g/mL at 25°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title