missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Chiralplate™ for TLC Enantiomer Separation
Resolve a wide variety of enantiomers. Chiralplate™ for TLC Enantiomer Separation consists of a special layer available on glass plates with fluorescent indicator.
£85.55 - £760.00
Specifications
Type | Glass Plate |
---|---|
Fluorescent Indicator | UV254 |
For Use With (Application) | For TLC Enantiomer Separation |
Thickness (Metric) | 0.25 mm |
Product Code | Brand | Dimensions (L x W) | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Quantity | Price | Quantity & Availability | ||||
10422052
|
Macherey-Nagel™
811058 |
10 x 10 cm | 25 |
£760.00
Pack of 25 |
Estimated Shipment: 11-09-2024
Log in to see stock. |
||||
12770184
|
Macherey-Nagel™
811059 |
10 x 20 cm | 4 |
£350.00
Pack of 25 |
Estimated Shipment: 11-09-2024
Log in to see stock. |
||||
10428781
|
Macherey-Nagel™
811057 |
5 x 20 cm | 50 |
£478.00
Pack of 50 |
Estimated Shipment: 11-09-2024
Log in to see stock. |
||||
12740184
|
Macherey-Nagel™
811056 |
10 x 20 cm | 4 Pk. |
£85.55
Pack of 4 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
10786322
|
Macherey-Nagel™
811055 |
10 x 20 cm | 25 Pk. |
£432.00
Pack of 25 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
Description
Features- Reversed phase nano silica impregnated with Cu(II) ions and a chiral selector (a proline derivative, DP 31 43 726 and EP 0 143 147)
- Separation based on ligand exchange, i. e. formation of ternary mixed-ligand complexes with the Cu(II) ions; differences in the stability of the diastereomeric complexes cause chromatographic separation
- Available as glass plates with fluorescent indicator
Application
- Enantiomer separation of amino acids, N-methylamino acids, N-formylamino acids, α-alkylamino acids, thiazolidine derivatives, dipeptides, lactones, α-hydroxycarboxylic acids
- A review on the application of CHIRALPLATE has been given by K. Günther [J. Chromatogr. 448 (1988) 11–30].
Specifications
Glass Plate | |
UV254 | |
For TLC Enantiomer Separation | |
0.25 mm |