missing translation for 'onlineSavingsMsg'
Learn More
Silicon(IV) chloride, 99.8+%, Thermo Scientific Chemicals Product Code.: 10606412

Silicon(IV) chloride, 99.8+%, Thermo Scientific Chemicals

Product Code. 10606412
500 mL, Glass bottle
Click to view available options
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10606412

Brand: Thermo Scientific Acros 201165000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-04-7
Molecular Formula Cl4Si
Molecular Weight (g/mol) 169.89
MDL Number MFCD00011229
InChI Key FDNAPBUWERUEDA-UHFFFAOYSA-N
Synonym silicon tetrachloride, silicon chloride, perchlorosilane, silane, tetrachloro, silicon iv chloride, tetrachlorosilicon, chlorid kremicity, silicio tetracloruro di, siliciumtetrachlorid, silicon chloride sicl4Show MoreShow Less
PubChem CID 24816
IUPAC Name tetrachlorosilane
SMILES Cl[Si](Cl)(Cl)Cl

Specifications

Boiling Point 58.0°C
Melting Point -70.0°C
Color Colorless
Physical Form Liquid
Quantity 500 mL
Assay Percent Range 99.8+%
Linear Formula SiCl4
Fieser 10,347; 11,466; 16,317
Merck Index 15, 8639
Solubility Information Solubility in water: decomposes. Other solubilities: miscible with benzene, ether, chloroform,, petroleum ether, carbon tetrachloride,, tin tetrachloride, sulfur, mono- and dichlorides, decomposes by alcohol with evolution of hydrogen, chloride
Formula Weight 169.9
Percent Purity ≥99.8%
Packaging Glass bottle
Refractive Index 1.413
Specific Gravity 1.48
Density 1.4800g/mL
Chemical Name or Material Silicon(IV) chloride, 99.8+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0415051 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number201165000,201160500
Lot NumberA0458105 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number201165000,201160500
Lot NumberA0466110 Certificate TypeCertificate of Analysis Date12/09/2024 Catalog Number201165000,201160500
Lot NumberA0466109 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog Number201165000,201160500
Lot NumberA0466114 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog Number201165000,201160500
compliance-icons compliance-icons
Product Identifier
  • Silicon(IV) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Silicon(IV) chloride, 99.8+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.