Learn More
Silicone oil, high temperature, usable temperature range: 25 to 250°C (open system) and 25 to 315°C (closed system), Thermo Scientific Chemicals
£63.32 - £613.05
Chemical Identifiers
CAS | 68083-14-7 |
---|---|
Molecular Formula | C16H22O2Si2 |
Molecular Weight (g/mol) | 302.52 |
MDL Number | MFCD00132673 |
InChI Key | ARWRSWALIGRKQA-UHFFFAOYSA-N |
Synonym | diphenyl-dimethylsiloxane copolymer, polydimethyl-diphenylsiloxane, viscosity 400cst. |
PubChem CID | 121233058 |
IUPAC Name | methoxy-dimethyl-[methyl(diphenyl)silyl]oxysilane |
SMILES | CO[Si](C)(C)O[Si](C)(C1=CC=CC=C1)C2=CC=CC=C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15435005
|
Thermo Scientific Alfa Aesar
045896.22 |
100 g |
£63.32
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15445005
|
Thermo Scientific Alfa Aesar
045896.36 |
500 g |
£199.64
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15455005
|
Thermo Scientific Alfa Aesar
045896.A4 |
2.5 kg |
£613.05
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Silicone oil is used in high temperature open systems, closed systems and circulating, closed loop and heat transfer baths. It is used as a high temperature fluid for laboratory research apparatus and instruments. It is also used as a calibration media in temperature controlled baths.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- High temperature open system, closed system, circulating, closed loop, and heat transfer baths
- High temperature fluids for laboratory research apparatus and instruments
Chemical Identifiers
68083-14-7 | |
302.52 | |
ARWRSWALIGRKQA-UHFFFAOYSA-N | |
121233058 | |
CO[Si](C)(C)O[Si](C)(C1=CC=CC=C1)C2=CC=CC=C2 |
C16H22O2Si2 | |
MFCD00132673 | |
diphenyl-dimethylsiloxane copolymer, polydimethyl-diphenylsiloxane, viscosity 400cst.Show More | |
methoxy-dimethyl-[methyl(diphenyl)silyl]oxysilane |
Specifications
68083-14-7 | |
Liquid | |
MFCD00132673 | |
14,8495 | |
Miscible with aliphatic,aromatic hydrocarbons,alcohol and chlorinated hydrocarbons. Immiscible with water,methanol and ethylene glycol. | |
CO[Si](C)(C)O[Si](C)(C1=CC=CC=C1)C2=CC=CC=C2 | |
302.52 | |
1.505 | |
Silicone oil, High temperature, Usable temp. range: 25 to 250°C (open system) and 25 to 315°C (closed system) |
Colorless | |
C16H22O2Si2 | |
>300°C (572°F) | |
diphenyl-dimethylsiloxane copolymer, polydimethyl-diphenylsiloxane, viscosity 400cst. | |
ARWRSWALIGRKQA-UHFFFAOYSA-N | |
methoxy-dimethyl-[methyl(diphenyl)silyl]oxysilane | |
121233058 | |
1.07 g/mL |
Safety and Handling
missing translation for 'rtecsNumber' : VW3146875
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.