missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Silicone sealing ring, for GL threads, without PTFE washer

Seal for screw connection cap.

Brand:  DWK Life Sciences 286200602

Code : NEW

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 10345191

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Suitable for PBT screw cap. Exemplary application: Fixing tubes

  • Suitable for PBT screw cap
Specifications

Specifications

6 mm
10/Pk.
16 mm
Fixing tubes; Sealing
Silicone
GL 18
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers