Learn More
DWK Life Sciences Silicone sealing ring, for GL threads, without PTFE washer
Seal for screw connection cap.
£1.15 - £63.45
Specifications
Material | Silicone |
---|---|
For Use With (Application) | Fixing tubes; Sealing |
Quantity | 10/Pk. |
Product Code | Brand | Diameter (Metric) Inside | Joint Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Joint Size | Price | Quantity & Availability | ||||
15931541
|
DWK Life Sciences
286200405 |
6 mm | GL 14 |
£19.61
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
15874598
|
DWK Life Sciences
286201007 |
10 mm | GL 18 |
£14.70
Pack of 10 |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||||
15369797
|
DWK Life Sciences
286201204 |
8 mm | GL 25 |
£23.15
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
10548411
|
DWK Life Sciences
286204604 |
26 mm | GL 45 |
£4.53
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||||
15824618
|
DWK Life Sciences
286218007 |
14 mm | GL 32 |
£34.95
Pack of 10 |
Estimated Shipment: 19-09-2024
Log in to see stock. |
||||
15814618
|
DWK Life Sciences
286217808 |
12 mm | GL 32 |
£34.95
Pack of 10 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||||
15884598
|
DWK Life Sciences
286201607 |
12 mm | GL 25 |
£23.15
Pack of 10 |
Estimated Shipment: 19-09-2024
Log in to see stock. |
||||
15804618
|
DWK Life Sciences
286217602 |
10 mm | GL 32 |
£34.95
Pack of 10 |
Estimated Shipment: 19-09-2024
Log in to see stock. |
||||
15844618
|
DWK Life Sciences
286218401 |
18 mm | GL 32 |
£34.95
Pack of 10 |
Estimated Shipment: 19-09-2024
Log in to see stock. |
||||
15834618
|
DWK Life Sciences
286218204 |
16 mm | GL 32 |
£34.95
Pack of 10 |
Estimated Shipment: 19-09-2024
Log in to see stock. |
||||
15854618
|
DWK Life Sciences
286218607 |
20 mm | GL 32 |
£34.95
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
15217458
|
DWK Life Sciences
286204801 |
32 mm | GL 45 |
£63.45
Pack of 10 |
Estimated Shipment: 19-09-2024
Log in to see stock. |
||||
10141831
|
DWK Life Sciences
286200808 |
8 mm | GL 18 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10345191
|
DWK Life Sciences
286200602 |
6 mm | GL 18 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10420303
|
DWK Life Sciences
286201401 |
10 mm | GL 25 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Suitable for PBT screw cap. Exemplary application: Fixing tubes
- Suitable for PBT screw cap
Specifications
Silicone | |
Fixing tubes; Sealing | |
10/Pk. |