missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ SILICOSEN™ Silicone Caps

Excellent permeability and secure fitting, thanks to seal lip. Ideally suitable for agitated cultures.

Brand:  Hirschmann™ 8905755

Code : A0

Additional Details : Weight : 0.26100kg

 View more versions of this product

Product Code. 11745524

  • £198.00 / Pack of 10
Estimated Shipment: 31-07-2024
to see stock.
Add to basket

Description

Description

  • Uniform pore structure
  • Good air permeability
  • Chemically and temperature resistant
  • Liquid repellent and reusable
Specifications

Specifications

Pink
For The Preparation And Sterilization Of Culture Media
10/Pk.
28 mm
Silicone Stopper
40 to 55 mm
Silicone
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers