Learn More
Silver cyanide, 99%, Thermo Scientific Chemicals
Silver plating
£96.08 - £303.51
Chemical Identifiers
CAS | 506-64-9 |
---|---|
Molecular Formula | CAgN |
Molecular Weight (g/mol) | 133.886 |
MDL Number | MFCD00003409 |
InChI Key | LFAGQMCIGQNPJG-UHFFFAOYSA-N |
Synonym | silver cyanide, silver cyanide ag cn, silver 1+ cyanide, kyanid stribrny czech, cyanure d'argent french, rcra waste number p104, silver i cyanide, rcra waste no. p104, kyanid stribrny, cyanure d'argent |
PubChem CID | 10475 |
IUPAC Name | silver;cyanide |
SMILES | [C-]#N.[Ag+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11371249
|
Thermo Scientific Alfa Aesar
011424.14 |
25 g |
£96.08
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361249
|
Thermo Scientific Alfa Aesar
011424.22 |
100 g |
£303.51
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Silver cyanide is used in silver-plating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
506-64-9 | |
133.886 | |
LFAGQMCIGQNPJG-UHFFFAOYSA-N | |
10475 | |
[C-]#N.[Ag+] |
CAgN | |
MFCD00003409 | |
silver cyanide, silver cyanide ag cn, silver 1+ cyanide, kyanid stribrny czech, cyanure d'argent french, rcra waste number p104, silver i cyanide, rcra waste no. p104, kyanid stribrny, cyanure d'argent | |
silver;cyanide |
Specifications
320°C (decomposition) | |
White | |
CAgN | |
MFCD00003409 | |
14,8512 | |
Insoluble in water,dilute acids,and alcohol; Soluble in alkali cyanides and boiling concentrated HNO3 | |
[C-]#N.[Ag+] | |
133.886 | |
133.84 | |
Odorless | |
Light sensitive | |
Silver cyanide |
506-64-9 | |
Powder | |
AgCN | |
UN1684 | |
silver cyanide, silver cyanide ag cn, silver 1+ cyanide, kyanid stribrny czech, cyanure d'argent french, rcra waste number p104, silver i cyanide, rcra waste no. p104, kyanid stribrny, cyanure d'argent | |
LFAGQMCIGQNPJG-UHFFFAOYSA-N | |
silver;cyanide | |
10475 | |
99% | |
1.685 | |
3.95 g/mL |
Safety and Handling
GHS H Statement
H300-H310-H330-H318-H290-H315
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
May be corrosive to metals.
Causes skin irritation.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
H300+H310+H330
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SILVER CYANIDE
missing translation for 'einecsNumber' : 208-048-6
missing translation for 'rtecsNumber' : VW3850000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.