missing translation for 'onlineSavingsMsg'
Learn More

Silver sulfate, pure, 10 g/l solution in sulfuric acid, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 279480025

Code : 29

Additional Details : CAS Number : 10294-26-5 Weight : 4.60000kg Transport : UN number : 1830 Chem class : 8 Pack group : II


 View more versions of this product

Product Code. 10247460

  • £231.00 / 2.50 L
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Silver sulfate
10g/L Solution in Sulfuric Acid
Ag2O4S
MFCD00003407
1.8400g/mL
silver sulfate, disilver sulfate, disilver 1+ sulfate, unii-8qg6hv4zpo, disilver 1+ sulphate, sulfuric acid, disilver 1+ salt, 8qg6hv4zpo, disilver 1+ ion sulfate, sulfuric acid, silver 1+ salt 1:2, sulfuric acid disilver i salt
[O-]S(=O)(=O)[O-].[Ag+].[Ag+]
311.792
311.79
Glass bottle
2.5 L
Colorless
10294-26-5,7664-93-9
Ag2SO4
01,1015; 03,254; 04,435
15, 8668
YPNVIBVEFVRZPJ-UHFFFAOYSA-L
disilver;sulfate
159865
Pure
1.84
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Silver sulfate, 10 g/l solution in sulfuric acid

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only